Learn More
Selleck Chemical LLC Zanamivir S3007-1g
SDP
Supplier:  Selleck Chemical LLC S30071G
Encompass_Preferred
Zanamivir (GG167 GR 121167X) is a neuraminidase inhibitor used in the treatment and prophylaxis of influenza caused by influenza A virus and influenza B virus
Catalog No. 50-311-1298