Learn More
Selleck Chemical LLC -Zanubrutinib E2654-5MG
SDP
Supplier:  Selleck Chemical LLC E26545MG
Encompass_Preferred
( )-Zanubrutinib (( )-BGB-3111) is the racemate of Zanubrutinib (HY-101474A) ( )-Zanubrutinib inhibits Bruton s tyrosine kinase (Btk) with an IC50 of 0 63 nM
Catalog No. 50-000-40315