Learn More
Medchemexpress LLC (±)-Zanubrutinib | 1633350-06-7 | 99.4% | 471.55 | C27H29N5O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101474100MG
Encompass_Preferred
(±)-Zanubrutinib is the racemate of zanubrutinib and a potent, selective inhibitor of Bruton's tyrosine kinase (Btk) with a reported IC50 of 0.63 nM. Supplied as research-grade solid or DMSO solution in multiple sizes for biochemical and cellular studies.
  • High purity (~99.4%) suitable for research.
  • Molecular weight 471.55; formula C27H29N5O3.
  • Available as solid or 10 mM solution in DMSO, multiple package sizes including 100 MG.
  • Potent Btk inhibition for biochemical and cellular assays.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-48223