Learn More
Medchemexpress LLC Zanubrutinib-d5 | 00-00-0 | 99.0% | 476.58 | C27H24D5N5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101474S5MG
Encompass_Preferred
Zanubrutinib-d5 is a deuterium-labeled isotopologue of zanubrutinib supplied as a research-grade reference standard for analytical, pharmacokinetic, and mechanistic studies. The compound incorporates five deuterium atoms (C27H24D5N5O3) and is provided as a high-purity powder suitable for LC-MS and tracer experiments.
  • Deuterium-labeled isotopologue for tracer and quantification studies.
  • Suitable for LC-MS internal standard and pharmacokinetic assays.
  • High purity for improved analytical accuracy.
  • Stable powder formulation with specified storage conditions.
  • Available in small mg pack sizes for method development and validation.
Catalog No. 50-002-48808