Learn More
Medchemexpress LLC HY-136179 5mg Medchemexpress, ZAP-180013 CAS:873080-25-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1361795MG
Encompass_Preferred
Medchemexpress, HY-136179 5mg ZAP-180013 CAS:873080-25-2 ZAP-180013 is a zeta-chain-associated protein kinase 70 (ZAP-70) inhibitor with an IC50 of 1.8 μM. ZAP-180013 inhibits the interaction of ZAP-70 SH2 domain with immunoreceptor tyrosine-based activation motif (ITAMs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0404