Learn More
Medchemexpress LLC Zapalog | 1708091-24-0 | 99.6% | 1108.24 g·mol⁻¹ | C58H73N7O15 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12631625MG
Encompass_Preferred
Zapalog is a photocleavable small-molecule heterodimerizer used to reversibly control protein-protein interactions. It dimerizes proteins tagged with FKBP and DHFR domains and is cleaved by light to rapidly terminate interactions, enabling temporal control in chemical biology and optogenetics experiments.
  • Photocleavable heterodimerizer allows rapid, reversible control of protein interactions.
  • Compatible with FKBP and DHFR tagged proteins for inducible dimerization.
  • Enables precise temporal control via light-induced photolysis.
  • Supplied as a research-grade small molecule with high purity.
  • Soluble in DMSO for preparation of working solutions.
Catalog No. 50-002-36298