Learn More
Medchemexpress LLC Zastaprazan | 2133852-18-1 | 100.0% | 362.47 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139557
Encompass_Preferred
Zastaprazan is a proton pump inhibitor and a potent potassium-competitive acid blocker. It is used in the research of gastrointestinal inflammatory diseases or gastric acid-related diseases, such as gastroesophageal reflux disease.
  • Acts as a proton pump inhibitor
  • Potent potassium-competitive acid blocker
  • Researched for gastrointestinal inflammatory diseases
  • Researched for gastric acid-related diseases
  • Researched for gastroesophageal reflux disease
Catalog No. 50-003-49948