Learn More
TARGETMOL CHEMICALS INC Zavondemstat 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T701211MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Zavondemstat (QC8222 free base) is a histone lysine demethylase 4 (KDM4) inhibitor with anticancer and antitumor activity for the study of triple-negative and breast cancers. Purity 99.78%
Catalog No. 50-287-7906