Learn More
Medchemexpress LLC ZB-R-55 | 2416593-55-8 | 99.4% | 441.48 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1610591MG
Encompass_Preferred
ZB-R-55 is an orally active RIPK1 inhibitor that can be used for sepsis study. It demonstrates anti-necroptotic activity in human U-937 cells, inhibiting LPS/Z-VAD-fmk induced necroptosis with an IC50 of 0.34 nM.
  • Orally active RIPK1 inhibitor.
  • Applicable for sepsis studies.
  • Exhibits anti-necroptotic activity.
Catalog No. 50-003-94537