Learn More
Aviva Systems Biology Corp ZBTB33 Antibody - N-terminal region (ARP38864_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38864_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZBTB33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZBTB33. Alternate names: ZNF348, ZNF-kaisoApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56825