Learn More
Medchemexpress LLC ZCL278 | 587841-73-4 | 98.5% | 584.89 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1396350MG
Encompass_Preferred
ZCL278 is a selective Cdc42 inhibitor with Kds of 6.4 μM in fluorescence titration and 11.4 μM in surface plasmon resonance (SPR) measurement. It can disrupt the Cdc42-ITSN interaction and GTP/GDP binding. ZCL278 shows inhibitory effects on various enveloped viruses but is ineffective against non-enveloped viruses. It is also used in studies of arsenic neurotoxicity and HER2-positive gastric cancer (GC).
  • Impedes wound healing in PC-3 cells
  • Inhibits Cdc42 activity in PC-3 cells
  • Inhibits mice primary neurons branching and growth cone motility
  • Prevents release of Junin virus (JUNV) ribonucleoprotein cores
  • Inhibits VSV, LCMV, and DV2 infections
  • Improves cytotoxicity of NaAsO2 in rat astrocytes
  • Reduces cholesterol levels and reverses collateral resistance in TC cells
  • Inhibits JUNV replication in a mouse model
  • Reduces cholesterol enrichment and reverses collateral resistance in vivo
Catalog No. 50-003-47915