Learn More
Medchemexpress LLC HY-U00002 5mg Medchemexpress, ZD-4190 CAS:413599-62-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU000025MG
Encompass_Preferred
Medchemexpress, HY-U00002 5mg ZD-4190 CAS:413599-62-9 ZD-4190 is a potent, orally available inhibitor of the vascular endothelial cell growth factor receptor 2 ( VEGFR2 ) and of epidermal growth factor receptor ( EGFR ) signalling, used for the treatment of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4403