Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zd7288 | 133059-99-1 | MFCD00914937 | 99.8% | 292.81 g/mol | C15H21ClN4 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY101346100MG

Encompass_Preferred

ZD7288 (ICI D7288) is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker used as a research reagent to inhibit HCN-mediated currents in electrophysiology and pharmacology studies. Supplied as a high-purity solid for in vitro research; not for human or clinical use.

  • Selective HCN channel blocker for in vitro electrophysiology.
  • High purity (~99.8%) suitable for research applications.
  • White to off-white solid form for convenient handling and storage.
  • Molecular weight 292.81 g/mol; formula C15H21ClN4.
  • Packaged as 100 MG for laboratory use.
  • Storage: 4°C; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-12998


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.