Learn More
Medchemexpress LLC α-Zearalenol | 36455-72-8 | 1 ML
Supplier: Medchemexpress LLC HYN671010MM/1ML
α-Zearalenol is a mycotoxin exhibiting high affinity for estrogen receptors (ER) and is a derivative of zearalenone (ZEN). It is known to cause reproductive disorders due to its xenoestrogenic effects. Available as a white to off-white solid, this product is intended for research use only.
- Exhibits high affinity for estrogen receptors (ER)
- Causes reproductive disorders due to xenoestrogenic effects
- Induces an increase in cell viability in MCF-7 cells (0.001-10 μM)
- Increases MCF7 cell population in S phase and G2/M phase
- Reduces expression of TNF-α, IL-1β, and IL-8 in HepG2 cells (1-10 μM)
- Soluble in DMSO at 100 mg/mL