Learn More
Medchemexpress LLC α-Zearalenol | 36455-72-8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN671010MG
Encompass_Preferred
Alpha-Zearalenol is a mycotoxin exhibiting high affinity for estrogen receptors (ER). This derivative of zearalenone (ZEN) is known for causing reproductive disorders in animals due to its xenoestrogenic effects. It is a solid, white to off-white compound with a molecular weight of 320.38 and a chemical formula of C18H24O5. This product is intended for research use only and not for patient administration.
  • Exhibits high affinity for estrogen receptors (ER)
  • Causes reproductive disorders due to xenoestrogenic effects
  • Inhibits cytotoxicity against human KB cells (IC50 100.29 μM)
  • Inhibits cytotoxicity against African green monkey Vero cells (IC50 60.33 μM)
  • Increases cell viability in MCF-7 cells
  • Reduces TNF-α expression
  • Induces cell cycle changes in MCF7 cells (S phase and G2/M phase)
Catalog No. 50-004-17990