Learn More
Zebra Technologies Corporation Zebra 105999-302 - Cleaning Card Kit
Supplier:  Zebra Technologies Corporation 105999302
Encompass_Preferred
Zebra 105999-302 Cleaning Card Kit 4 print engine cleaning cards and 4 feeder cleaning cards - Compatible with Zebra ZXP Series 3 card printer - 1,000 prints / card. This kit can be used with any ZXP3 printer (both Legacy and Enhanced versions)
Catalog No. NC3242221