Learn More
TRIBIOSCIENCE INC Zebrafish Fast Genotyping System
SDP
Supplier:  TRIBIOSCIENCE INC TBS4035
Encompass
The Fast Genotyping system is designed for rapid DNA extraction and PCR amplification from zebrafish tail or fin clip, or other tissue. The PCR product can be directly loaded onto gel for genotype analysis.
Catalog No. NC2580212