Learn More
Cayman Chemical ZebrafIsh GlucocortIcoId Rece
SDP
Supplier:  Cayman Chemical 325793X32 WELL
Encompass_Preferred
zfGR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. Z00201-32, Z00201]
Catalog No. 50-291-0733