Learn More
Selleck Chemical LLC Zebularine
SDP
Supplier:  Selleck Chemical LLC S711310MG
Encompass_Preferred
Zebularine (NSC 309132 4-Deoxyuridine) is a DNA methylation inhibitor that forms a covalent complex with DNA methyltransferases also inhibits cytidinedeaminase with Ki of 2 M in a cell-free assay
Catalog No. 50-136-4418