Learn More
Medchemexpress LLC Zederone | 7727-79-9 | 246.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN10505MG
Encompass_Preferred
Zederone is a sesquiterpene that inhibits ovarian cancer cell proliferation through the mTOR/p70s6K signalling pathway. It also inhibits CYP activities. Zederone demonstrates antibacterial activity against multi-drug resistant and Methicillin-resistant Staphylococcus aureus strains, shows cognition-improving capacity, and assists in modulating gut bacterial dysbiosis.
  • Inhibits ovarian cancer cell proliferation via the mTOR/p70s6K signalling pathway.
  • Shows antibacterial activity against multi-drug resistant and Methicillin-resistant Staphylococcus aureus strains.
  • Demonstrates cognition-improving capacity.
  • Assists in the modulation of gut bacterial dysbiosis.
  • Selectively kills leukemia stem cells (KG1a) with an IC50 of 56.8 μg/mL with low cytotoxicity towards normal peripheral blood mononuclear cells (PBMCs).
Catalog No. 50-004-17177