Learn More
Sigma Aldrich Fine Chemicals Biosciences Zein prolamine 1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z36251KG
Encompass
Zein is a prolamine and an alcohol soluble protein present in maize endosperm cells. Zein is a 20kDa structural protein and is encoded by the gene mapped on the short arm of maize chromosome 7. This protein lacks essential amino acids such as lysine and tryptophan. Therefore it has a poor nutritional value.
Catalog No. 50-290-2738