Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation ZenFit Tubing, 75 μm ID, 26 in.

Supplier:  Waters Corp 700011505

Encompass_Preferred

This ZenFit tubing is designed to connect the micro binary solvent manager (μBSM) to the column within an ACQUITY UPLC M-Class System setup. It features specific dimensions for precise applications.

  • ZenFit tubing type
  • Dimensions: 75 μm ID x 26 in.
  • Application: connects the micro binary solvent manager (μBSM) to the column
  • System compatibility: used within an ACQUITY UPLC M-Class System for LC conditions

Catalog No. 50-203-9845


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.