Learn More
Medchemexpress LLC Zenidolol hydrochloride | 72795-01-8 | MFCD01529928 | 99.7% | 313.86 g/mol | C17H28ClNO2 | 5 MG

Supplier: Medchemexpress LLC HY139515MG
Zenidolol hydrochloride is the hydrochloride salt of ICI-118551, a selective β2-adrenergic receptor antagonist used in pharmacology research to probe β-adrenergic receptor subtype function. The compound is supplied as a solid with high purity and documented physicochemical and solubility properties suitable for in vitro studies.
- CAS number 72795-01-8.
- Ki values: β2 0.7 nM, β1 49.5 nM, β3 611 nM.
- Purity 99.74%.
- Molecular weight 313.86 g/mol.
- Appearance solid, white to light yellow.
- Solubility DMSO 25 mg/mL; water 8.33 mg/mL (requires ultrasonic).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.