Learn More
Enterprise Technology Solutions Zep Commercial Cleaner/Degreaser, Concentrated, 5 Gallon, Purple
SDP
Supplier:  Enterprise Technology Solutions ZPE85635
Encompass_Preferred
Industrial cleaner/degreaser features a concentrated, alkaline formula to quickly remove a wide variety of greasy and carbonaceous soils not readily penetrated by ordinary cleaning products. It contains no phosphates to ensure safe, long-term use. It cleans industrial concrete floors and other masonry surfaces, greasy engines and equipment. Easy-to-use design can be applied with a mop, brush, low-pressure spray or automatic scrubbing machine. Versatile cleaner/degreaser is perfect for industrial cleaning applications, including carbon, soot, ink and grease removal.
Catalog No. 50-008-10495