Learn More
Enterprise Technology Solutions Zep Commercial Deodorizer,Water-Based,Blue Sky Scent,CL
SDP
Supplier:  Enterprise Technology Solutions ZPE203901
Encompass_Preferred
Air Fair Blue Sky is an environmentally friendly, water-based deodorant that contains a concentrated, highly effective blend of odor-neutralizing and masking agents. All-purpose design eliminates odors from stale cigarettes, cigar smoke, body wastes, food spillage, garbage, cooked foods, perspiration, pets and fire damage. Formula imparts a nice, fresh fragrance to the treated area. Ready-to-use spray does not require dilution. Spray bottle with trigger provides easy handling and doesn't cause waste or spills. Just a small amount delivers effective odor control and keeps working after several days or more in certain applications. Noncorrosive spray contains no phosphates or carcinogens and is made for industrial and business use only.
Catalog No. 50-008-10580