Learn More
Medchemexpress LLC Zetomipzomib maleate | 10MG
SDP
Supplier:  Medchemexpress LLC HY114419A10MG
Encompass_Preferred
Zetomipzomib (KZR-616) maleate a first-in-class immunoproteasome inhibitor selectively targets the LMP7 (IC50 39/57 nM hLMP7/mLMP7) and LMP2 (IC50 131/179 nM hLMP2/mLMP2) subunits of the immunoproteasome Zetomipzomib maleate has the potential for the research of multiple autoimmune diseases[1][2]
Catalog No. 50-002-30018