Learn More
Medchemexpress LLC HY-114419A 5mg , Zetomipzomib (maleate) CAS:2170983-62-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114419A5MG
Encompass_Preferred
HY-114419A 5mg Zetomipzomib (maleate) CAS: 2170983-62-5 Zetomipzomib (KZR-616) maleate, a first-in-class immunoproteasome inhibitor, selectively targets the LMP7 (IC50: 39/57 nM=hLMP7/mLMP7) and LMP2 (IC50: 131/179 nM=hLMP7/mLMP7) subunits of the immunoproteasome. Zetomipzomib maleate has the potential for the research of multiple autoimmune diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1301