Learn More
Electron Microscopy Sciences Z-Fix Concentrate, 1/EA
SDP
Supplier:  Electron Microscopy Sciences 6412501
Encompass_Preferred
Concetrate for bufferred aqueous zinc formalin. Zinc formalin fixatives improve general morphology and immunohistochemical staining and can reduce or eliminate the need for antigen recovery procedures while preserving immunoreactivity for months. Compatible with all tissue processors. Use in place of neutral bufferred formalin. Easy to dilute with water. 5 gal fixative with one gal concentrate.
Catalog No. 50-209-0860