Learn More
Medchemexpress LLC HY-120859 10mg , Zidebactam CAS:1436861-97-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12085910MG
Encompass_Preferred
HY-120859 10mg Zidebactam CAS: 1436861-97-0 Zidebactam (WCK-5107) is a potent β-lactamase inhibitor. Zidebactam also is a penicillin-binding protein2 (PBP2) inhibitor with an IC50 of 0.26 μg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0604