Learn More
Medchemexpress LLC Zidesamtinib | 2739829-00-4 | C22H22FN7O | 50 MG

Supplier: Medchemexpress LLC HY15229250MG
Zidesamtinib (NVL-520) is a potent, selective, orally active, and brain-penetrant inhibitor of diverse ROS1 fusions and resistance mutations, with IC50s of 0.7 and 7.9 nM for wild-type ROS1 and ROS1 G2032R, respectively. It spares TRK inhibition and can be used for cancer research.
- Potent, selective, orally active, and brain-penetrant inhibitor
- Inhibits diverse ROS1 fusions and resistance mutations
- Spares TRK inhibition
- Suitable for cancer research
- Purity of 99.78%
- Molecular weight: 419.45
- Solid, off-white to light yellow appearance
- Inhibits wild-type ROS1 fusions with average IC50s of 0.4 nM
- Inhibits ROS1 fusions with G2032R mutation with average IC50s of 1.6 nM
- Potently inhibits non-G2032R ROS1 mutants with IC50s ≤ 1.5 nM
- Induces tumor regression in wild-type ROS1 xenograft models
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.