Learn More
Medchemexpress LLC Zika virus E/envelope protein | 50 UG
SDP
Supplier:  Medchemexpress LLC HYP7445650UG
Encompass_Preferred
Recombinant Zika virus envelope (E) protein fragment supplied as a filtered protein solution for research use. The protein is expressed in HEK293 cells with a C-terminal His tag and is derived from the SPH2015 strain; formulation is in PBS, pH 7.4.
Catalog No. 50-002-94039