Learn More
Medchemexpress LLC Zika virus NS1 protein (HEK293, His) | >95.0% | 5 UG
SDP
Supplier:  Medchemexpress LLC HYP744555UG
Encompass_Preferred
Recombinant Zika virus nonstructural protein 1 (NS1) expressed in HEK293 human cells with an N-terminal His tag. Supplied as a purified protein reagent for use in immunoassays, antibody production, serology controls, western blotting, and assay development. Typical purity is ≥95% and molecular weight is approximately 50 kDa, provided in a buffered solution at a reported concentration.
Catalog No. 50-002-93838