Learn More
ATCC Zika virus; Strain: MR 766; Quantitative RNA
Supplier:  ATCC VR1838DQ
Encompass
Quantitative Genomic RNA from Zika virus strain MR 766 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-3886