Learn More
Apexbio Technology LLC Zileuton, 100mg. Cas: 111406-87-2 MFCD: MFCD00866097. 5-lipoxygenase (5-LOX) inhibitor, orally active
SDP
Supplier:  Apexbio Technology LLC A5384100
Encompass_Preferred
IC50: Zileuton inhibited 5-hydroxyeicosatetraenoic aicd synthesis in rat basophilic leukemia cell and polymorphonuclear leukocytes (IC50 = 0.5 and 0.3 M), respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4951