Learn More
Medchemexpress LLC Zileuton | 111406-87-2 | MFCD00866097 | 99.6% | 236.29 g/mol | C11H12N2O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY141645MG
Encompass_Preferred
Zileuton is a potent, selective inhibitor of 5-lipoxygenase used in research to study leukotriene-mediated inflammation and asthma-related pathways. Supplied as a small-quantity, high-purity research reagent, it is suitable for in vitro and in vivo studies that require modulation of the 5-lipoxygenase pathway.
  • Potent and selective 5-lipoxygenase inhibition.
  • High purity suitable for analytical and biological assays.
  • Available in small quantities for research use.
  • Useful for studies of leukotriene-mediated inflammation and asthma models.
  • COA and data sheet available to support quality control.
Catalog No. 50-002-21348