Learn More
Medchemexpress LLC Zileuton | 111406-87-2 | MFCD00866097 | 99.6% | C11H12N2O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14164100MG
Encompass_Preferred
Zileuton is a potent and selective inhibitor of 5-lipoxygenase with antiasthmatic properties, intended for research use only.
  • Potent and selective inhibitor of 5-lipoxygenase
  • Has antiasthmatic properties
  • Available as a solid
  • Melting point is 157-158°C
  • Stable under recommended storage conditions
  • Identified for laboratory chemical uses
Catalog No. 50-003-53705