Learn More
TARGETMOL CHEMICALS INC Zileuton 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0477200MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Zileuton (A 64077) is a synthetic derivative of hydroxyurea with antiasthmatic properties. The leukotriene inhibitor zileuton blocks 5-lipoxygenase, which catalyzes the formation of leukotrienes from arachidonic acid; causes bronchodilation; decreases bronchial mucous secretion and edema; and may prevent or decrease the symptoms of asthma. Purity 99.56%
Catalog No. 50-288-0548