Learn More
Medchemexpress LLC Zileuton sodium | 118569-21-4 | 98.0% | C11H11N2NaO2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14164A
Encompass_Preferred
Zileuton sodium is a potent and selective inhibitor of 5-lipoxygenase, exhibiting inflammatory activities. It is for research use only and not sold to patients.
  • Potent and selective 5-lipoxygenase inhibitor
  • Exhibits inflammatory activities
  • White to off-white solid appearance
  • High purity (98.02%)
  • Suitable for research use only
Catalog No. 50-003-52798