Learn More
Medchemexpress LLC Zileuton sodium | 118569-21-4 | 98.6% | 258.27 g/mol | C11H11N2NaO2S | 5 MG

Supplier: Medchemexpress LLC HY14164A5MG
Zileuton sodium is the sodium salt of zileuton, a potent and selective inhibitor of 5-lipoxygenase used as a research reagent to modulate leukotriene-mediated inflammatory pathways. It is supplied as a solid, high-purity chemical intended for in vitro and in vivo experimental use; not for human or therapeutic use.
- Potent, selective 5-lipoxygenase inhibitor
- Sodium salt form for improved solubility
- High purity (98.6%) suitable for research applications
- Molecular weight 258.27 g/mol; formula C11H11N2NaO2S
- Stable when stored sealed, away from moisture
- Provided in small research quantities appropriate for preclinical studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.