Learn More
Medchemexpress LLC Zim | 301298-87-3 | C20H19N3O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY152246
Encompass_Preferred
Zim is a norbornene derived from 4-aminoantipyrine, functioning as a potent inducer of DNA damage. It causes genomic and chromosomal damage, triggers cell death, and activates phagocytosis, indicating its chemotherapeutic potential for cancer research.
  • Potent inducer of DNA damage
  • Causes genomic and chromosomal damage
  • Induces cell death
  • Activates phagocytosis
  • Shows chemotherapeutic potential for cancer research
  • Reduces the frequency of chromosome micronucleus
  • Reduces cisplatin and doxorubicin-induced liver and kidney cell death
Catalog No. 50-003-74348