Learn More
Medchemexpress LLC ZIM | 301298-87-3 | C20H19N3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15224650MG
Encompass_Preferred
ZIM, a norbornene derived from 4-Aminoantipyrine, is a potent inducer of DNA damage, causing genomic and chromosomal damage as well as inducing cell death and activating phagocytosis. ZIM has chemotherapeutic potential for use in cancer research.
  • Potent inducer of DNA damage
  • Causes genomic and chromosomal damage
  • Induces cell death
  • Activates phagocytosis
  • Chemotherapeutic potential for use in cancer research
  • Can effectively reduce the frequency of chromosome micronucleus in adult male Swiss mice
  • Reduces the frequency of cisplatin-CIS and doxorubicin-DOX-induced liver and kidney cell death
Catalog No. 50-003-72889