Learn More
Medchemexpress LLC Zimelidine dihydrochloride | 60525-15-7 | 98.1% | 390.15 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY110023
Encompass_Preferred
Zimelidine dihydrochloride is a potent and selective inhibitor of serotonin 5-HT uptake and SERT. It is also an antidepressant.
  • Potent and selective inhibitor of serotonin 5-HT uptake and SERT.
  • Functions as an antidepressant.
  • For research use only.
  • Inhibits serotonin reuptake.
  • Affects neurotransmission.
Catalog No. 50-003-47357