Learn More
Strem, An Ascensus Company CAS# 136-53-8. 100g. Zinc 2-ethylhexanoate, 99% (~18% Zn, 1% Diethylene glycol monomethyl ether). MFCD00053316
SDP
Supplier:  Strem, An Ascensus Company 303038100G
Encompass
CAS# 136-53-8. 100g. Zinc 2-ethylhexanoate, 99% (~18% Zn, 1% Diethylene glycol monomethyl ether). MFCD00053316. Molecular Weight: 351.79. Molecular Formula: Zn[OOCCH(C2H5)C4H9]2. Color/form: slightly yellow viscous liq. Strem# 30-3038. http://www.strem.com/catalog/v/30-3038/
Catalog No. 50-901-11390