Learn More
Medchemexpress LLC Zinc acetate | 557-34-6 | 99.1% | 10 G

Supplier: Medchemexpress LLC HYB181710G
Zinc acetate is a biological molecule for research use only. It is presented as a white to off-white solid with a molecular weight of 183.47. This product is suitable for various laboratory applications.
- Molecular formula: Zn(CH3CO2)2
- Appearance: White to off-white solid
- Purity: 99.05%
- Storage: Stable at room temperature for 3 years; in solvent, stable for 2 years at -80°C or 1 year at -20°C
- pH: 6.35
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.