Learn More
Strem, An Ascensus Company CAS# 5263-02-5. 100g. Zinc carbonate hydroxide. MFCD00011519
SDP
Supplier:  Strem, An Ascensus Company 933005100G
Encompass
CAS# 5263-02-5. 100g. Zinc carbonate hydroxide. MFCD00011519. Molecular Weight: 548.92. Molecular Formula: 3Zn(OH)2∙2ZnCO3. Color/form: white pwdr. Strem# 93-3005. http://www.strem.com/catalog/v/93-3005/
Catalog No. 50-901-14957