Learn More
Hach Company Zinc Chemkey Reagents
Supplier:  Hach Company 9879000
Encompass
Zinc Chemkey Reagents (box of 25)For determination of Zinc using Hach Method 10313Ensure SL1000/SL250 has been updated with Software Version V3 0 12 1489 to use this Chemkey
Catalog No. 50-002-07029