Learn More
Eurofins Discoverx Zinc Chloride, 10 mg. (92-1213)
Supplier:  Eurofins Discoverx 921213
Encompass
Zinc Chloride is an ionic water-soluble compound with wide applications, including textile processing, metallurgical fluxes, and chemical synthesis like Fischer Indole synthesis and the Friedel-Crafts acylation reactions. Also used in cosmetics such as dentifrices, mouthwash, skin care, hair conditioners, and bath products. Acts as an agonist for GPR39. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9738