Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Granular, USP, 97-100.5%, Spectrum™ Chemical
SDP

ZI105, 7646-85-7, ZnCl2

$336.50 - $3680.00

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18-607-840
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
ZI105100GM
100 g Amber Glass Bottle
Each for $336.50
Add to Cart
 
18-607-841
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
ZI105500GM
500 g Amber Glass Bottle
Each for $497.50
Add to Cart
 
18-607-842
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
ZI1052.5KG
2.5 kg Amber Glass Bottle
Each for $1,434.00
Add to Cart
 
18-607-843
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
ZI10512KG
12 kg Amber Glass Bottle
Each for $3,680.00
Add to Cart
 
Description

Description

Spectrum™ Chemical Zinc Chloride, Granular, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L

Specifications

7646-85-7
Cl2Zn
136.28
USP
1
JIAARYAFYJHUJI-UHFFFAOYSA-L
97 to 100.5%
Zinc Chloride
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.