Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Granular, USP, 97-100.5%, Spectrum™ Chemical
SDP

ZI105, 7646-85-7, ZnCl2

Supplier:  Spectrum Chemical Manufacturing Corporation ZI105500GM

Quantity 500 g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18-607-841


Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart

Description

Description

Spectrum™ Chemical Zinc Chloride, Granular, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7646-85-7
Cl2Zn
136.28
JIAARYAFYJHUJI-UHFFFAOYSA-L

Specifications

7646-85-7
Cl2Zn
136.28
USP
500 g
1
JIAARYAFYJHUJI-UHFFFAOYSA-L
97 to 100.5%
Amber Glass Bottle
Zinc Chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $500 in Chemicals with Purchase
Spend $250 or more on Spectrum Chemical chemicals and get up to $500 to spend on additional chemicals.

Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.