Learn More
Sigma Aldrich Fine Chemicals Biosciences Zinc chloride puriss. meet500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 14422500G
Encompass
Zinc chloride is a zinc salt. Zinc chloride in combination with 1-ethyl-3-methylimidazolium chloride comprises a molten salt which is employed in the electrodeposition studies of zinc.Zinc chloride is a colorless white compound composed of zinc and chlorine which can be used as a flux for soldering.
Catalog No. 50-289-9660